Order Entry
Sweden
ContactUsLinkComponent
Automatic isoperibol calorimeter, Model 6400
Automatic isoperibol calorimeter, Model 6400
Catalog # PARR6400EF
Supplier:  PARR
undefined
Automatic isoperibol calorimeter, Model 6400
Catalog # PARR6400EF
Supplier:  PARR

Specifications

  • Description:
    Model 6400 automatic isoperibol calorimeter
  • W×D×H:
    420×460×510 mm
  • Pk:
    1
  • Type:
    Isoperibol, water jacket

Specifications

About this item

The 6400 automatic isoperibol calorimeter represents the next evolutionary step in the Parr automated calorimeters.

  • Automatic oxygen fill
  • Automatic bucket fill
  • Automatic bomb wash
  • Parr fixed bomb and bucket technology offer unique levels of automation for the entire calorimetric determination

Designed for laboratories that require high through-put, the 6400 automatic isoperibol calorimeter features a closed loop cooling subsystem and the highest automation in a calorimeter. The patented Quick Twist-Lock vessel closure design allows one operator to run up to four 6400 calorimeters simultaneously.

Precision: 0,1% Class
Test frequency: 6 to 7/h

Ordering information: For more information on options, accessories and other models please contact your Avantor represenatative.
Delivery: 6400 automatic isoperibol calorimeter with 1138 (CL) oxygen combustion vessel, A570DD oxygen regulator, A812DD nitrogen regulator, A1589DD rinse tank, and 6038 500 firings kit.