Sulpho-SMPB (Sulphosuccinimidyl 4-(4-maleimidophenyl)butyrate)
Catalog # BC24
Supplier: G-Biosciences
CAS Number:
Specifications
- Pk:100 mg
- Seq:2440
Specifications
Specification Test Results
- Spacer Arm (Å):14.5
- Reactive Toward:Primary Amine + Sulfhydryl
- Membrane Permeable:No
- Water Soluble:Yes
- Cleavable/ Reversible:No
Specification Test Results
About this item
:
:458.374 g/mol
CAS Number:92921-26-1
Formula:C₁₈H₁₅N₂O₉S·Na
MDL Number:MFCD00054981