Order Entry
Sweden
ContactUsLinkComponent
Sulpho-KMUS, Pierce™
Sulpho-KMUS, Pierce™
Catalog # PIER21111
CAS Number:  
undefined
Sulpho-KMUS, Pierce™
Catalog # PIER21111
CAS Number:  

Specifications

  • Förp.:
    50 mg
  • Seq:
    2370

Specifications

About this item

 :
 :480.46 g/mol
Storage Temperature:Kylskåp
CAS Number:211236-68-9
Formula:C₁₉H₂₅N₂NaO₉S
MDL Number:MFCD01863412