Mildronate dihydrate
Catalog # ACRO466260010
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
Specifications
Specification Test Results
- Appearance:White to beige crystalline powder
- Infrared spectrum:Conforms
- Titration with HClO4:97.5 to 102.5%
- Water:18 to 22%
Specification Test Results
About this item
:
:182.22 g/mol
Melting Pt:85°C…90°C
:GHS07
Storage Temperature:Refrigerator
CAS Number:86426-17-7
Formula:C₆H₁₄N₂O₂·₂H₂O
MDL Number:MFCD00876389