Anti-ARID5A Mouse Monoclonal Antibody [clone: 1683]
Catalog # NOVUNBP2-43676
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen symbol:ARID5A
- Clonality:Monoclonal
- Clone:1683
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human
- Western blot:Yes
- Gene ID:10865
- Antigen synonyms:AT-rich interactive domain-containing protein 5A|ARID domain-containing protein 5A|AT rich interactive domain 5A (MRF1-like)|RFVG5814
- Storage buffer:PBS
- Storage temperature:Aliquot and store at -20 °C or -80 °C. Avoid freeze-thaw cycles.
- Concentration:2,14 mg/ml
- Immunogen:Recombinant protein encompassing a sequence within the center region of human ARID5A. The exact sequence is proprietary.
- Purification:Protein A purified
- Pk:0,1 mL
Specifications
About this item
The ARID5A antibody (1683) from Novus Biologicals is a mouse monoclonal antibody to ARID5A. This antibody reacts with human. The ARID5A antibody (1683) has been validated for the following applications: Western Blot.
Type: Primary
Antigen: ARID5A
Clonality: Monoclonal
Clone: 1683
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human