Anti-ALDH1L1 Mouse Monoclonal Antibody [clone: 4A12]
Catalog # NOVUNBP2-50045SS
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen symbol:ALDH1L1
- Clonality:Monoclonal
- Clone:4A12
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG2b
- Reactivity:Pig,Horse,Chicken,Human,Bovine,Rat,Mouse
- Western blot:Yes
- Gene ID:10840
- Antigen synonyms:FDH|member L1|aldehyde dehydrogenase 1 family|FTHFD10-fTHF|EC 1.5.1.6,10-FTHFDH|aldehyde dehydrogenase family 1 member L1|DKFZp781N0997|Cytosolic 10-formyltetrahydrofolate dehydrogenase|10-formyltetrahydrofolate dehydrogenase|formyltetrahydrofolate dehydrogenase
- Storage buffer:PBS
- Storage temperature:Store at 4 °C short term. Aliquot and store at -20 °C long term. Avoid freeze-thaw cycles.
- Concentration:1 mg/ml
- Immunogen:Amino acids 1- 401 of human ALDH1L1. [UniProt# O75891]
- Purification:Immunogen affinity purified
- Pk:25 µl
Specifications
About this item
The ALDH1L1 Antibody (4A12) from Novus Biologicals is a mouse monoclonal antibody to ALDH1L1. This antibody reacts with human, mouse, rat, porcine, bovine, chicken, equine. The ALDH1L1 Antibody (4A12) has been validated for the following applications: western blot, immunocytochemistry / immunofluorescence.
Type: Primary
Antigen: ALDH1L1
Clonality: Monoclonal
Clone: 4A12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat, Porcine, Bovine, Chicken, Equine