Anti-CNPase chicken polyclonal antibody
NOVUNB100-1935
- Antibody type:Primary
- Antigen symbol:CNPase
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Chicken
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Reactivity:Mouse
- Western blot:Yes
- Gene ID:1267
- Antigen synonyms:EC 3.1.4.37|3' cyclic nucleotide 3' phosphohydrolase|2'-3'-cyclic-nucleotide 3'-phosphodiesterase|2'-3'-cyclic nucleotide 3' phosphodiesterase|2'|CNP1|CNPase
- Storage buffer:10 mM Sodium PBS (0,9% isotonic, w/v, pH 7,2)
- Storage temperature:Store at 4 °C, do not freeze.
- Concentration:0,1 mg/ml
- Immunogen:Chickens were immunized with synthetic peptide/ keyhole limpet hemocyanin (KLH) conjugate. The sequence of the synthetic peptide was shared between the human (NP_149124, NCBI) and mouse (P16330, NCBI) sequences. After repeated injections into the hens, immune eggs were collected, and the IgY fractions were purified from the yolks. These IgY fractions were then affinity-purified using a peptide column, the concentrations of the eluate adjusted to 100 ug/ml, and the preparation was filter-sterilised.
- Purification:Immunogen affinity purified
- Size:0,25 ml
- Pk:0,25 mL
The CNPase Antibody from Novus Biologicals is a chicken polyclonal antibody to CNPase. This antibody reacts with mouse. The CNPase Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.