Order Entry
Sweden
Orders LinkContactUsLinkComponent
Human recombinant STING M284 variant (from <i>E. coli</i>)

Human recombinant STING M284 variant (from E. coli)

100 µG
Catalog # CAYM23594-100
Supplier:   Cayman Chemical
Human recombinant STING M284 variant (from <i>E. coli</i>)
Catalog # CAYM23594-100
Supplier:  Cayman Chemical
CAS Number:  
Human recombinant STING M284 variant (from <i>E. coli</i>)
Human recombinant STING M284 variant (from E. coli)

100 µG

CAS Number:  
Price
14 475,00 kr
Log in for availability
1 * 100 µG
Product Details & Documents

An active, pure human recombinant enzyme.

STING M284 variant (human recombinant) contains amino acids 138-379 of the wild-type variant (R232) with a methionine substituted for arginine at position 284. Stimulator of interferon genes (STING) is a component of the innate immune response that binds to cyclic dinucleotides, which are bacterial second messengers, leading to activation of NF-κB and transcription of immunomodulatory genes, including type I interferon (IFN). The R232 variant of STING is the most common variant in the human population, found at a frequency of 57,9% in the 1000 Genome Project. The SNP variant H232 is found at a 13,7% frequency. The R284M mutation in STING is associated with constitutive activation of downstream signalling. It increases the propensity of STING to dimerize and associate with the kinase TBK1, enhancing the ability of STING to activate IRF3 and NF-κB and induce a type I IFN response. However, the R284M mutation occurs outside of the dimerization region between positions 153-177, so rather than a direct effect on dimerization, it is predicted to promote or inhibit binding of a cellular factor that stabilizes or impairs STING dimerization.

Used for: Infectious Disease, Autoimmunity

Endotoxin level and content: Not tested

Specifications

Specifications

  • Pk:
    100 µG
  • Cat. No.:
    CAYM23594-100
  • Enzyme type:
    Recombinant
  • Storage conditions:
    –80 °C
  • Enzyme synonyms:
    Endoplasmic Reticulum Interferon Stimulator, ERIS, hSTING, MITA, Stimulator of Interferon Genes, TMEM173, Transmembrane Protein 173
  • Formulation:
    50 mM HEPES pH 8,0, 150 mM NaCl, 10% glycerol
  • Purity:
    ≥80% estimated by SDS-PAGE
  • Species:
    Human
  • Source:
    E. coli
  • Enzyme name:
    STING M284 variant
  • Molecular weight:
    28,8 kDa
  • Shipping conditions:
    –80 °C, dry ice

Specifications