Human recombinant frataxin (from E. coli)
Catalog # NOVUNBP1-30312
Supplier: Novus Biologicals
CAS Number:
Specifications
- Pk:0,1 mg
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Source:E. coli
- Species:Human
- Storage conditions:Store at 4 °C short term. Aliquot and store at –20 °C long term. Avoid freeze-thaw cycles.
- Gene ID:2395
- Protein synonyms:FARRmitochondrial|Friedreich ataxia protein|EC 1.16.3.1|frataxin|CyaY|Friedreich ataxia|FA|Fxn|X25MGC57199
- Protein/peptide name:Frataxin
- Purity:>95% pure by SDS-PAGE
- Concentration:1 mg/ml
- Tested applications:ELISA
Specifications
About this item
The Recombinant Human Frataxin Protein from Novus Biologicals is derived from E. coli. The Recombinant Human Frataxin Protein has been validated for the following applications: SDS-Page.