Anti-MBD1 Mouse monoclonal antibody [clone: 100B272.1]
Catalog # NOVUNB100-56537SS
Supplier: Novus Biologicals
Specifications
- Antibody type:Primary
- Antigen symbol:MBD1
- Clonality:Monoclonal
- Clone:100B272.1
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human,Mouse
- Western blot:Yes
- Gene ID:4152
- Antigen synonyms:the regulator of fibroblast growth factor 2 (FGF-2) transcription|PCM1CXXC3RFT|Methyl-CpG-binding protein MBD1|methyl-CpG-binding domain protein 1|CXXC-type zinc finger protein 3|methyl-CpG binding domain protein 1 isoform PCM1|Protein containing methyl-CpG-binding domain 1|methyl-CpG binding domain protein 1
- Storage buffer:PBS containing 0.05% BSA
- Storage temperature:Store at 4 °C short term. Aliquot and store at −20 °C long term. Avoid freeze-thaw cycles.
- Concentration:0,5 mg/ml
- Immunogen:This antibody was generated by immunizing mice with a synthetic peptide corresponding to amino acids 391-405 (SESEDGAGSPPPYRR) of human MBD1; GenBank no ref NP_056671.2
- Purification:Protein G purified
- Pk:25 µG
Specifications
About this item
The MBD1 Antibody (100B272.1) from Novus Biologicals is a mouse monoclonal antibody to MBD1. This antibody reacts with human, mouse. The MBD1 Antibody (100B272.1) has been validated for the following applications: Western Blot, Chromatin Immunoprecipitation, Immunoprecipitation.
Type: Primary
Antigen: MBD1
Clonality: Monoclonal
Clone: 100B272.1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse