Anti-ATG5 Chicken Polyclonal Antibody
NOVUNBP1-76992
- Antibody type:Primary
- Antigen name:Autophagy Related 5
- Antigen symbol:ATG5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Chicken
- ImmunoChemistry:Yes
- Isotype:IgY
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Gene ID:9474
- Antigen synonyms:ASPAPG5-LIKE|Autophagy protein 5|APG5LS. cerevisiae)-like|ATG5 autophagy related 5 homolog (S. cerevisiae)|APG5 autophagy 5-like (S. cerevisiae)
- Storage buffer:PBS
- Storage temperature:Store at 4 °C short term. Aliquot and store at −20 °C long term. Avoid freeze-thaw cycles.
- Concentration:1,0 mg/ml
- Immunogen:Antibody was raised against a 16 amino acid synthetic peptide from near the amino terminus of human ATG5. The immunogen is located within the first 50 amino acids of ATG5.
- Purification:Immunogen affinity purified
- Size:100 μg
- Pk:0,1 mg
The ATG5 Antibody from Novus Biologicals is a chicken polyclonal antibody to ATG5. This antibody reacts with human, mouse, rat. The ATG5 Antibody has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Recommended dilutions: Immunohistochemistry (parafin): 2,5 µg/ml.
Type: Primary
Antigen: ATG5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human, Mouse, Rat