GF Signet Installation Tee Fittings, CPVC/PVC 80
Supplier: Avantor Fluid Handling
Certificates
About this item
Easily install your flowmeters.
- Constructed of durable CPVC/PVC 80
PVC Tee Fittings for PVC 80 pipes up to 4" in diameter and CPVC 80 pipes up to 1 1/2" in diameter. Slip on ends (no threads) sensors.
- Environmentally Preferable:
Specifications
- Connection type:
- 1/2" ID Slip Fit
- 1" ID Slip Fit
- 1-1/2" ID Slip Fit
- 2" ID Slip Fit
- 4" ID Slip Fit
- 1" ID Slip Fit
- Warranty:
- 1 year part and 2 years labor
- 1 year part and 2 years labor
- 1 year part and 2 years labor
- 1 year part and 2 years labor
- 1 year part and 2 years labor
- 1 year part and 2 years labor