Order Entry
Sweden
ContactUsLinkComponent
Orthophosphoric acid 85%, CMOS for microelectronic, J.T.Baker®
Corrosive
Orthophosphoric acid 85%, CMOS for microelectronic, J.T.Baker®
Catalog # 0264-35
Supplier:  Avantor
CAS Number:  
Corrosive
Orthophosphoric acid 85%, CMOS for microelectronic, J.T.Baker®
Catalog # 0264-35
Supplier:  Avantor
CAS Number:  

Specifications

  • Pk:
    5 L
  • Seq:
    5190

Specifications

Specification Test Results

  • Assay (H3PO4):
    85.0 - 87.0 %
  • Chloride (Cl):
    max. 1 ppm
  • Color (APHA):
    max. 10
  • Color after Heating:
    passes test
  • Heavy Metals (as Pb):
    max. 5 ppm
  • Nitrate (NO3):
    max. 2 ppm
  • Particles > 0.5 µm/ml:
    max. 150
  • Particles > 1.0 µm/ml:
    max. 10
  • Reducing Substances:
    passes test
  • Specific Gravity at 60°/60°F:
    1.691 - 1.710
  • Sulfate (SO4):
    max. 12 ppm
  • Volatile Acids:
    max. 0.16 ueq/g
  • Aluminium (Al):
    max. 0.5
  • Antimony (Sb):
    max. 10
  • Arsenic (As):
    max. 0.05
  • Calcium (Ca):
    max. 1.5
  • Chromium (Cr):
    max. 0.2
  • Cobalt (Co):
    max. 0.05
  • Copper (Cu):
    max. 0.05
  • Gold (Au):
    max. 0.3
  • Iron (Fe):
    max. 2
  • Lead (Pb):
    max. 0.3
  • Lithium (Li):
    max. 0.1
  • Magnesium (Mg):
    max. 0.2
  • Manganese (Mn):
    max. 0.1
  • Nickel (Ni):
    max. 0.2
  • Potassium (K):
    max. 1.5
  • Sodium (Na):
    max. 2.5
  • Strontium (Sr):
    max. 0.1
  • Titanium (Ti):
    max. 0.3
  • Zinc (Zn):
    max. 2

Specification Test Results

About this item

 :
 :98 g/mol
Melting Pt:28 °C
Density:1.71…1.87 g/cm³ (25 °C)
Storage Temperature:Ambient
EINECS:231-633-2
UN:1805
ADR:8,III
 :GHS05
CAS Number:7664-38-2
Formula:H₃PO₄
MDL Number:MFCD00011340