AP-conjugate stabiliser, SeramunStab® STAP
APOSBIL3116-100ML
:
- Pk:100 mL
- Description:SeramunStab® STAP AP-Conjugate stabiliser
- Seq:01
Liquid product colourless odourless pH-value 5,9 to 6,1, Stabiliser for very low concentrated AP conjugates. Suitable for proteins resistant to pH values <6,0.