Mark 7 spray chamber supplies
Supplier: VARIAN
Certificates
About this item
Mark 7 Spray Chambers for flame atomisation System. Easy-to-use twist and lock assembly allows for easy cleaning and routine maintenance.
- Inert construction makes spray chamber suitable for use with aqueous and organic solutions
- Fit organic resistant O-rings to ensure organic compatibility
Flexible operation with the capability to tune performance using the externally adjustable impact bead, and to fit mixing paddles for better precision, or both.
Delivery: Mark 7 operating supplies kit available to support routine operation. Contents: Venturi nebuliser (PEEK), nebuliser capillary kit, nebuliser block, glass impact beads, high solids capillary tubing, O-ring kit for aqueous samples, mixing paddles, burner cleaning strips.