Candida antarctica Lipase A
Supplier: Apollo Scientific
Certificates
About this item
Covalent on IB-150A. Activity: 2000 TBU/g
- immobilized on Immobead 150
Specifications
- Species:Candida antarctica
- Storage conditions:Refrigerator
- Enzyme name:Lipase A