Order Entry
Sweden
ContactUsLinkComponent
Anti-CD8 Mouse Antibody (Biotin)
Anti-CD8 Mouse Antibody (Biotin)
Catalog # APOSBITP2010-1MG
Supplier:  Apollo Scientific
Anti-CD8 Mouse Antibody (Biotin)
Catalog # APOSBITP2010-1MG
Supplier:  Apollo Scientific

Specifications

  • Antigen symbol:
    CD8
  • Conjugation:
    Biotin
  • Host:
    Mouse
  • Reactivity:
    Human
  • Pk:
    1 mg

Specifications

About this item