Gå till Rubrik
Gå till Huvudinnehåll
Gå till Sidfot
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Proteins and Peptides
Recombinant Rubella Virus E1 Mosaic
Recombinant Rubella Virus E1 Mosaic
Recombinant Rubella Virus E1 Mosaic
Catalog # APOSBITP1936-1MG
Supplier:
Apollo Scientific
CAS Number:
Expand image
Recombinant Rubella Virus E1 Mosaic
Catalog # APOSBITP1936-1MG
Supplier:
Apollo Scientific
CAS Number:
Specifications
Pk:
1 mg
Protein/peptide type:
Recombinant
Protein synonyms:
Rubella Mosaic
Protein/peptide name:
Rubella Virus E1 Mosaic
Specifications
About this item
Product Details
Product Details