Gå till Rubrik
Gå till Huvudinnehåll
Gå till Sidfot
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Proteins and Peptides
Enfuvirtide T-20
Enfuvirtide T-20
Enfuvirtide T-20
Catalog # APOSBITP1540-100MG
Supplier:
Apollo Scientific
CAS Number:
Expand image
Enfuvirtide T-20
Catalog # APOSBITP1540-100MG
Supplier:
Apollo Scientific
CAS Number:
Specifications
Pk:
100 mg
Protein synonyms:
Enfuvirtide
Protein/peptide name:
Enfuvirtide T-20
Specifications
About this item
Product Details
Product Details