Order Entry
Sweden
ContactUsLinkComponent
 
Human Recombinant IGFBP-1
Catalog # APOSBITP1111-5UG
Leverantör:  Apollo Scientific
CAS-nummer:  
Human Recombinant IGFBP-1
Catalog # APOSBITP1111-5UG
Leverantör:  Apollo Scientific
CAS-nummer:  

Specifikationer

  • Pk:
    5 µG
  • Protein/peptide type:
    Recombinant
  • Species:
    Human
  • Protein synonyms:
    Insulin Like Growth Factor Binding Protein-1
  • Protein/peptide name:
    IGFBP-1

Specifikationer

Om denna produkt