Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3,4,5-Trifluorobenzaldehyde
3,4,5-Trifluorobenzaldehyde 98%
3,4,5-Trifluorobenzaldehyde 98%
:
Apollo Scientific
3,4,5-Trifluorobenzaldehyde 98%
:
Apollo Scientific
:
132123-54-7
:
:
C₇H₃F₃O
:
MFCD00083523
:
160.1 g/mol
:
174 °C
:
24…26 °C
:
1.42 g/cm³ (25 °C)
:
63 °C
:
GHS07
:
Ambient