
4-Fluor-3-(isopropylcarbamoyl)phenylboronic acid 98%
Catalog # APOSPC5031-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:225.03 g/mol
Melting Pt:263…265 °C
:GHS07
Storage Temperature:Ambient
CAS Number:874219-21-3
Formula:C₁₀H₁₃BFNO₃
MDL Number:MFCD08235040