Gå till Rubrik
Gå till Huvudinnehåll
Gå till Sidfot
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Magnesium fluoride
Magnesiumfluorid, pulver
Magnesiumfluorid, pulver
:
Apollo Scientific
Magnesiumfluorid, pulver
:
Apollo Scientific
:
7783-40-6
:
:
MgF₂
:
MFCD00011108
:
62.3 g/mol
:
2239 °C
:
1261 °C
:
3,148 g/cm³ (25 °C)
:
Omgivande
:
231-995-1
:
GHS07