Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4,4,5,5,6,6,6-Heptafluoro-1-hexanol
4,4,5,5,6,6,6-Heptafluoro-1-hexanol
4,4,5,5,6,6,6-Heptafluoro-1-hexanol
Irriterande
4,4,5,5,6,6,6-Heptafluoro-1-hexanol
Leverantör:
Apollo Scientific
Certifikat
Expand image
Irriterande
4,4,5,5,6,6,6-Heptafluoro-1-hexanol
Leverantör:
Apollo Scientific
Om denna produkt
CAS-nummer:
679-02-7
:
Formel:
C₆H₇F₇O
MDL-nummer:
MFCD00077554
:
228.11 g/mol
Kokande Pt:
140…141 °C/745mmHg
:
GHS07
Förvaringstemperatur:
Ambient