Order Entry
Sweden
ContactUsLinkComponent
2-Chloro-5-fluorophenol 98%
Corrosive
Irritant
Environment
2-Chloro-5-fluorophenol 98%
Catalog # APOSPC1880A-5G
Supplier:  Apollo Scientific
CAS Number:  
Corrosive
Irritant
Environment
2-Chloro-5-fluorophenol 98%
Catalog # APOSPC1880A-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :146.55 g/mol
Boiling Pt:183…185 °C
Storage Temperature:Ambient
UN:3265
ADR:8,III
 :GHS05
 :GHS07
 :GHS09
CAS Number:3827-49-4
Formula:C₆H₄ClFO
MDL Number:MFCD00042524