Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Hem
4-Bromotetrafluoroaniline
4-Bromotetrafluoroaniline 98%
4-Bromotetrafluoroaniline 98%
4-Bromotetrafluoroaniline 98%
Leverantör:
Apollo Scientific
Certifikat
Expand image
4-Bromotetrafluoroaniline 98%
Leverantör:
Apollo Scientific
Om denna produkt
CAS-nummer:
1998-66-9
:
Formel:
C₆H₂BrF₄N
MDL-nummer:
MFCD00012144
:
243.9844 g/mol
Smält Pt:
59…61 °C
:
GHS07
Förvaringstemperatur:
Refrigerator