Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,3-Difluoro-4-toluoyl chloride
2,3-Difluoro-4-toluoyl chloride
2,3-Difluoro-4-toluoyl chloride
Corrosive
2,3-Difluoro-4-toluoyl chloride
Supplier:
Apollo Scientific
Certificates
Expand image
Corrosive
2,3-Difluoro-4-toluoyl chloride
Supplier:
Apollo Scientific
About this item
CAS Number:
261763-38-6
:
Formula:
C₈H₅ClF₂O
MDL Number:
MFCD01631645
ADR:
8,II
:
190.58 g/mol
UN:
3265
:
GHS05
Storage Temperature:
Ambient