Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-(4-Chlorophenyl)propionic acid
2-(4-Chlorophenyl)propionic acid 96%
2-(4-Chlorophenyl)propionic acid 96%
2-(4-Chlorophenyl)propionic acid 96%
Supplier:
Apollo Scientific
Certificates
Expand image
2-(4-Chlorophenyl)propionic acid 96%
Supplier:
Apollo Scientific
About this item
CAS Number:
938-95-4
:
Formula:
C₉H₉ClO₂
MDL Number:
MFCD00044670
:
184.62 g/mol
Storage Temperature:
Ambient