

3-Methylcyclohexylamine (cis- and trans- mixture) 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:6850-35-7
:
Formula:C₇H₁₅N
MDL Number:MFCD00001494
:113.20 g/mol
Boiling Pt:151 °C
Density:0.85 g/cm³ (25 °C)
Storage Temperature:Ambient
UN:2733
ADR:3+8,II
:GHS02
:GHS05