Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Allyl-p-tolyl ether
Allyl-p-tolyl ether 95%
Allyl-p-tolyl ether 95%
Allyl-p-tolyl ether 95%
Supplier:
Apollo Scientific
Certificates
Expand image
Allyl-p-tolyl ether 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
23431-48-3
:
Formula:
C₁₀H₁₂O
MDL Number:
MFCD00026098
:
148.21 g/mol
Storage Temperature:
Ambient