Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2'-Hydroxy-5'-methylpropiophenone
2'-Hydroxy-5'-methylpropiophenone 95%
2'-Hydroxy-5'-methylpropiophenone 95%
2'-Hydroxy-5'-methylpropiophenone 95%
Supplier:
Apollo Scientific
Certificates
Expand image
2'-Hydroxy-5'-methylpropiophenone 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
938-45-4
:
Formula:
C₁₀H₁₂O₂
MDL Number:
MFCD01098830
Storage Temperature:
Ambient