2-(5-Bromo-2-pyridylazo)-5-(diethylamino)phenol
Catalog # APOSOR921049-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:349.23 g/mol
Melting Pt:157…158 °C
Storage Temperature:Refrigerator
CAS Number:14337-53-2
Formula:C₁₅H₁₇BrN₄O
MDL Number:MFCD00006255