N-Acetyl-D-leucine 98%
Catalog # APOSOR912302-100G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:173.2 g/mol
Storage Temperature:Ambient
CAS Number:19764-30-8
Formula:C₈H₁₅NO₃
MDL Number:MFCD00066069