Resorcinol 99%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:108-46-3
:
Formula:C₆H₆O₂
MDL Number:MFCD00002269
Boiling Pt:281 °C
Melting Pt:109…111 °C
Density:1.272 g/cm³ (25 °C)
Flash Pt:127 °C
:110.11 g/mol
Storage Temperature:Ambient