Thiazole-5-carbonitrile 95%
Catalog # APOSOR905805-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:110.1371 g/mol
Storage Temperature:Ambient
CAS Number:25742-12-5
Formula:C₄H₂N₂S
MDL Number:MFCD06797206