Tri-tert-butyl-1,4,7,10-tetraazacyclododecane-1,4,7,10-tetraacetate 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:137076-54-1
:
Formula:C₂₈H₅₂N₄O₈
MDL Number:MFCD02259697
:572.74 g/mol
Melting Pt:88…93 °C (dec.)
Storage Temperature:Refrigerator