Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-Pyrimidin-2-yl-1H-pyrrole-2-carbaldehyde
1-Pyrimidin-2-yl-1H-pyrrole-2-carbaldehyde 95%
1-Pyrimidin-2-yl-1H-pyrrole-2-carbaldehyde 95%
1-Pyrimidin-2-yl-1H-pyrrole-2-carbaldehyde 95%
Supplier:
Apollo Scientific
Certificates
Expand image
1-Pyrimidin-2-yl-1H-pyrrole-2-carbaldehyde 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
383136-27-4
:
Formula:
C₉H₇N₃O
MDL Number:
MFCD02665163
:
173.18 g/mol
Storage Temperature:
Ambient