Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Chloro-2-methylbenzyl alcohol
4-Chloro-2-methylbenzyl alcohol 98%
4-Chloro-2-methylbenzyl alcohol 98%
4-Chloro-2-methylbenzyl alcohol 98%
Leverantör:
Apollo Scientific
Certifikat
Expand image
4-Chloro-2-methylbenzyl alcohol 98%
Leverantör:
Apollo Scientific
Om denna produkt
CAS-nummer:
129716-11-6
:
Formel:
C₈H₉ClO
MDL-nummer:
MFCD06201140
:
156.61 g/mol
Förvaringstemperatur:
Ambient