
5-Nitroacenaphthene, Technical Grade
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:602-87-9
:
Formula:C₁₂H₉NO₂
MDL Number:MFCD00014277
:199.21 g/mol
Melting Pt:100…102 °C
EINECS:210-025-0
:GHS08
Storage Temperature:Ambient