Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Butynyl toluene-4-sulphonate
3-Butynyl toluene-4-sulphonate
3-Butynyl toluene-4-sulphonate
3-Butynyl toluene-4-sulphonate
Supplier:
Apollo Scientific
Certificates
Expand image
3-Butynyl toluene-4-sulphonate
Supplier:
Apollo Scientific
About this item
CAS Number:
23418-85-1
:
Formula:
C₁₁H₁₂O₃S
MDL Number:
MFCD00041687
:
224.28 g/mol
Storage Temperature:
Ambient