Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Beställningsinmatning
Orderstatus
Ditt konto
Varukorg
Sweden
Beställningsinmatning
Orderstatus
Ditt konto
Varukorg
ContactUsLinkComponent
Hem
2'-Formylbiphenyl-4-carboxylic acid
2'-Formylbiphenyl-4-carboxylic acid
2'-Formylbiphenyl-4-carboxylic acid
:
Apollo Scientific
2'-Formylbiphenyl-4-carboxylic acid
:
Apollo Scientific
:
205871-49-4
:
:
C₁₄H₁₀O₃
:
MFCD03424615
:
GHS07
:
Ambient
:
226.2300 g/mol