0,00 kr
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Sweden
ContactUsLinkComponent
2-Ethoxybenzamide
2-Ethoxybenzamide
2-Ethoxybenzamide
APOSOR70170-500G
:
Apollo Scientific
:
2-Ethoxybenzamide
APOSOR70170-500G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
500 g
Seq:
3645
:
:
165.19 g/mol
:
132…134 °C
:
GHS07
:
Ambient
:
938-73-8
:
C₉H₁₁NO₂
:
MFCD00007977