Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4,5-Dichloro-2-nitroaniline
4,5-Dichloro-2-nitroaniline 97%
4,5-Dichloro-2-nitroaniline 97%
:
Apollo Scientific
4,5-Dichloro-2-nitroaniline 97%
:
Apollo Scientific
:
6641-64-1
:
:
C₆H₄Cl₂N₂O₂
:
MFCD00007770
:
207.02 g/mol
:
177…182 °C
:
229-657-3
:
GHS07
:
Ambient