Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
(1S)-(-)-Camphanic chloride
(1S)-(-)-Camphanic chloride
(1S)-(-)-Camphanic chloride
(1S)-(-)-Camphanic chloride
Supplier:
Apollo Scientific
Certificates
Expand image
(1S)-(-)-Camphanic chloride
Supplier:
Apollo Scientific
About this item
CAS Number:
39637-74-6
:
Formula:
C₁₀H₁₃ClO₃
MDL Number:
MFCD00135626
:
216.67 g/mol
Melting Pt:
69…71 °C
Storage Temperature:
Ambient