Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2,4,6-Trimethylbenzoyl chloride
2,4,6-Trimethylbenzoyl chloride
2,4,6-Trimethylbenzoyl chloride
:
Apollo Scientific
2,4,6-Trimethylbenzoyl chloride
:
Apollo Scientific
:
938-18-1
:
:
C₁₀H₁₁ClO
:
MFCD00013650
:
182.65 g/mol
:
143…146 °C/60mmHg
:
1.095 g/cm³ (25 °C)
:
>110 °C
:
Refrigerator
:
213-339-6
:
3265
:
8,III
:
GHS05