Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Methoxy-3-methylbenzaldehyde
4-Methoxy-3-methylbenzaldehyde
4-Methoxy-3-methylbenzaldehyde
4-Methoxy-3-methylbenzaldehyde
APOSOR60083-25G
:
Apollo Scientific
:
4-Methoxy-3-methylbenzaldehyde
APOSOR60083-25G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
25 g
Seq:
3308
:
:
150.18 g/mol
:
80…85 °C/1mmHg
:
1.025 g/cm³ (25 °C)
:
112 °C
:
251-177-8
:
GHS07
:
Ambient
:
32723-67-4
:
C₉H₁₀O₂
:
MFCD00003375