0,00 kr
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Sweden
ContactUsLinkComponent
cis-2-Butene
cis-2-Butene
cis-2-Butene
:
Apollo Scientific
cis-2-Butene
:
Apollo Scientific
:
590-18-1
:
:
C₄H₈
:
2.1,
:
56.11 g/mol
:
1012
:
GHS02
:
Ambient