

trans-1,4-Diaminocyclohexane 97%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2615-25-0
:
Formula:C₆H₁₄N₂
MDL Number:MFCD00075174
:114.19 g/mol
Boiling Pt:197 °C
Melting Pt:68…72 °C
Flash Pt:71 °C
Storage Temperature:Ambient
UN:3259
ADR:8,II
:GHS05
:GHS07