Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Beställningsinmatning
Orderstatus
Ditt konto
Varukorg
Sweden
Beställningsinmatning
Orderstatus
Ditt konto
Varukorg
ContactUsLinkComponent
Hem
Benzeneruthenium(II) chloride dimer
Benzeneruthenium(II) chloride dimer 95+%
Benzeneruthenium(II) chloride dimer 95+%
Benzeneruthenium(II) chloride dimer 95+%
Supplier:
Apollo Scientific
Certificates
Expand image
Benzeneruthenium(II) chloride dimer 95+%
Supplier:
Apollo Scientific
About this item
CAS Number:
37366-09-9
:
Formula:
C₁₂H₁₂Cl₄Ru₂
MDL Number:
MFCD00064686
:
500.17 g/mol
Melting Pt:
250 °C (dec.)
:
GHS07
Storage Temperature:
Ambient