5-Chloro-2-thiophenesulphonyl chloride 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:2766-74-7
:
Formula:C₄H₂Cl₂O₂S₂
MDL Number:MFCD00051667
:217.1 g/mol
Boiling Pt:112…117 °C/15mmHg
Melting Pt:26…28 °C
Density:1.623 g/cm³ (25 °C)
Storage Temperature:Refrigerator
UN:3261
ADR:8,III
:GHS05